Assay Details
| Gene Symbol: Taf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233151.2 | XP_006233213.1 | 1 - 2 | 100 | 77 | |
| GenBank mRNA | BC166866.1 | - | 1 - 2 | 64 | 8369659 | 77 |
| FM135814.1 | - | 1 - 2 | 38 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166866.1,XM_006233151.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf13
Genomic Map