Assay Details
Gene Symbol: Zdhhc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039339.1 | NP_001034428.1 | 2 - 3 | 526 | 55 | |
GenBank mRNA | AY886534.1 | - | 2 - 3 | 526 | 55 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
