Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025699.2 | NP_001020870.2 | 6 - 7 | 1333 | 76 | |
| XM_008764240.1 | XP_008762462.1 | 5 - 6 | 1529 | 76 | ||
| XM_008764241.1 | XP_008762463.1 | 6 - 7 | 1285 | 76 | ||
| GenBank mRNA | AY845021.1 | - | 6 - 7 | 1333 | 76 | |
| BC097999.1 | - | 6 - 7 | 1259 | 7453136 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem57
Genomic Map