Assay Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025699.2 | NP_001020870.2 | 7 - 8 | 1500 | 110 | |
| XM_008764240.1 | XP_008762462.1 | 6 - 7 | 1696 | 110 | ||
| XM_008764241.1 | XP_008762463.1 | 7 - 8 | 1452 | 110 | ||
| GenBank mRNA | AY845021.1 | - | 7 - 8 | 1500 | 110 | |
| BC097999.1 | - | 7 - 8 | 1426 | 7453136 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem57
Genomic Map