Assay Details
| Gene Symbol: Gja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021654.2 | NP_067686.1 | 1 - 2 | 58 | 71 | |
| GenBank mRNA | BC078837.1 | - | 1 - 2 | 58 | 7128770 | 71 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC078837.1,NM_021654.2 transcripts.
Set Membership: |
|

Gja4
Genomic Map