Assay Details
| Gene Symbol: Cda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108688.1 | NP_001102158.1 | 303 | 68 | ||
| Gene Symbol: LOC100909857 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750066.3 | XP_003750114.1 | 2 - 3 | 320 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cda
Genomic Map