Assay Details
| Gene Symbol: Hdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025409.1 | NP_001020580.1 | 5 - 6 | 518 | 72 | |
| GenBank mRNA | BC059156.1 | - | 5 - 6 | 535 | 72 | |
| BC097943.1 | - | 5 - 6 | 518 | 7458532 | 72 | |
| BC107476.1 | - | 5 - 6 | 515 | 7443793 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hdac1
Genomic Map