Assay Details
| Gene Symbol: Ajap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101014.1 | NP_001094484.1 | 3 - 4 | 1322 | 106 | |
| GenBank mRNA | AB179740.1 | - | 3 - 4 | 1322 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ajap1
Genomic Map