Assay Details
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115036.1 | NP_001108508.1 | 1 - 2 | 72 | 143 | |
GenBank mRNA | BC107929.1 | - | 1 - 2 | 78 | 143 | |
BC158610.1 | - | 1 - 2 | 51 | 7374180 | 143 | |
FQ225384.1 | - | 1 - 2 | 71 | 143 | ||
FQ232337.1 | - | 1 - 2 | 71 | 143 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC158610.1,NM_001115036.1 transcripts.
This assay is in the 5'UTR region of the BC158610.1,NM_001115036.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
