Assay Details
Gene Symbol: Taf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115036.1 | NP_001108508.1 | 4 - 5 | 519 | 141 | |
XM_006239067.2 | XP_006239129.1 | 5 - 6 | 638 | 141 | ||
XM_006239068.2 | XP_006239130.1 | 4 - 5 | 623 | 141 | ||
GenBank mRNA | BC107929.1 | - | 5 - 6 | 653 | 141 | |
BC158610.1 | - | 4 - 5 | 498 | 7374180 | 141 | |
FQ225384.1 | - | 4 - 5 | 518 | 141 | ||
FQ232337.1 | - | 4 - 5 | 518 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
