Assay Details
Gene Symbol: Pramel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109617.1 | NP_001103087.1 | 334 | 67 | ||
XM_006239342.2 | XP_006239404.1 | 2 - 3 | 477 | 67 | ||
XM_006239343.2 | XP_006239405.1 | 3 - 4 | 644 | 67 | ||
XM_008764290.1 | XP_008762512.1 | 486 | 67 |
Gene Symbol: LOC103692518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764326.1 | XP_008762548.1 | 1 - 2 | 519 | 67 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
