Assay Details
| Gene Symbol: Tmco4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034949.1 | NP_001030121.1 | 11 - 12 | 1407 | 67 | |
| XM_006239262.1 | XP_006239324.1 | 11 - 12 | 2832 | 67 | ||
| GenBank mRNA | BC099755.1 | - | 11 - 12 | 1407 | 5623720 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmco4
Genomic Map