Assay Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031598.3 | NP_113786.3 | 1 - 2 | 44 | 64 | |
| GenBank mRNA | FQ220554.1 | - | 1 - 2 | 44 | 64 | |
| FQ227448.1 | - | 1 - 2 | 44 | 64 | ||
| M25148.1 | - | 1 - 2 | 44 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the M25148.1, NM_031598.3 transcripts.
Set Membership: |
|

Pla2g2a
Genomic Map