Assay Details
| Gene Symbol: Fam132a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108000.1 | NP_001101470.1 | 1 - 2 | 426 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam132a| Gene Symbol: Fam132a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108000.1 | NP_001101470.1 | 1 - 2 | 426 | 94 | |
Set Membership: |
|