Assay Details
| Gene Symbol: Rap1gap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100713.1 | NP_001094183.1 | 7 - 8 | 506 | 64 | |
| XM_006239189.1 | XP_006239251.1 | 6 - 7 | 583 | 64 | ||
| XM_006239193.1 | XP_006239255.1 | 6 - 7 | 583 | 64 | ||
| GenBank mRNA | CB581996.1 | - | 7 - 8 | 518 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rap1gap
Genomic Map