Assay Details
| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106686.1 | NP_001100156.1 | 12 - 13 | 1684 | 78 | |
| XM_006238862.2 | XP_006238924.1 | 12 - 13 | 1956 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ago4
Genomic Map