Assay Details
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134689.1 | NP_001128161.1 | 4 - 5 | 596 | 71 | |
XM_008764171.1 | XP_008762393.1 | 4 - 5 | 3391 | 71 | ||
GenBank mRNA | BC168155.1 | - | 3 - 4 | 973 | 8362701 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
