Assay Details
| Gene Symbol: Ccdc28b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134689.1 | NP_001128161.1 | 5 - 6 | 623 | 69 | |
| XM_008764171.1 | XP_008762393.1 | 5 - 6 | 3418 | 69 | ||
| GenBank mRNA | BC168155.1 | - | 4 - 5 | 1000 | 8362701 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc28b
Genomic Map