Assay Details
| Gene Symbol: Syf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133417.1 | NP_596908.1 | 4 - 5 | 395 | 69 | |
| GenBank mRNA | AF366369.1 | - | 4 - 5 | 395 | 69 | |
| BC097289.1 | - | 4 - 5 | 403 | 7445525 | 69 | |
| FQ234066.1 | - | 2 - 3 | 222 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syf2
Genomic Map