Assay Details
Gene Symbol: Gpr153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034855.1 | NP_001030027.1 | 4 - 5 | 1129 | 85 | |
XM_006239485.2 | XP_006239547.1 | 4 - 5 | 3534 | 85 | ||
XM_006239486.2 | XP_006239548.1 | 4 - 5 | 3534 | 85 | ||
GenBank mRNA | BC105872.1 | - | 4 - 5 | 1129 | 7133996 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
