Assay Details
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 1879 | 106 |
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100538.1 | NP_001094008.1 | 11 - 12 | 1857 | 106 | |
XM_006238798.1 | XP_006238860.1 | 11 - 12 | 2445 | 106 | ||
XM_006238799.1 | XP_006238861.1 | 11 - 12 | 2445 | 106 | ||
GenBank mRNA | BC088337.1 | - | 6 - 7 | 804 | 106 | |
BC166452.1 | - | 11 - 12 | 1862 | 8372968 | 106 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
