Assay Details
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100538.1 | NP_001094008.1 | 1 - 2 | 563 | 73 | |
XM_006238798.1 | XP_006238860.1 | 1 - 2 | 1103 | 73 | ||
XM_006238799.1 | XP_006238861.1 | 1 - 2 | 1103 | 73 | ||
XM_006238800.1 | XP_006238862.1 | 1 - 2 | 1103 | 73 | ||
XM_006238801.1 | XP_006238863.1 | 1 - 2 | 1103 | 73 | ||
GenBank mRNA | BC166452.1 | - | 1 - 2 | 568 | 8372968 | 73 |
Gene Symbol: RGD1562451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769025.1 | XP_008767247.1 | 1 | 537 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
