Assay Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019202.2 | NP_062075.1 | 4 - 5 | 596 | 101 | |
| XM_006239142.2 | XP_006239204.1 | 4 - 5 | 614 | 101 | ||
| XM_006239143.2 | XP_006239205.1 | 3 - 4 | 375 | 101 | ||
| XM_008764212.1 | XP_008762434.1 | 4 - 5 | 689 | 101 | ||
| XM_008764213.1 | XP_008762435.1 | 4 - 5 | 467 | 101 | ||
| XM_008764214.1 | XP_008762436.1 | 4 - 5 | 2138 | 101 | ||
| GenBank mRNA | BC097325.1 | - | 4 - 5 | 596 | 7456563 | 101 |
| U07798.1 | - | 4 - 5 | 923 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pla2g2c
Genomic Map