Assay Details
| Gene Symbol: Plekhg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201272.1 | NP_958429.1 | 20 - 21 | 2612 | 73 | |
| GenBank mRNA | AY499658.1 | - | 20 - 21 | 2612 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P140238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140238 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140238 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P140238 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P540720 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63949 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5990 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10874275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874266 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10874258 | - | - |
| Rat Genome 230 2.0 Array | 1374697_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374697_at | - | - |
| Affymetrix HT Rat Focus Array | 1374697_at | - | - |
| Affymetrix HT Rat Focus Array | 1374697_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500014935.rn.1 | TC0500001994.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374697_at | - | - |
| Rat Expression Set 230 | 1374697_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10874258 | - | - |
| Rat Genome U34 Set | rc_AI044124_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21518761 | - | - |
| ABI Rat Genome Survey Microarray | 22413706 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365588 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49750 | - | - |

Plekhg5
Genomic Map