Assay Details
| Gene Symbol: Gpr137b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105978.1 | NP_001099448.1 | 4 - 5 | 570 | 127 | |
| XM_006254372.2 | XP_006254434.1 | 5 - 6 | 1072 | 127 | ||
| XM_006254373.2 | XP_006254435.1 | 5 - 6 | 1067 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr137b
Genomic Map