Assay Details
| Gene Symbol: Tmem185a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135712.1 | NP_001129184.1 | 2 - 3 | 377 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P597522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138655 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P597522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P597522 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P597522 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P499401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10935788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935787 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10935782 | - | - |
| Rat Genome 230 2.0 Array | 1376085_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376085_at | - | - |
| Affymetrix HT Rat Focus Array | 1376085_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376085_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800022406.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022405.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022411.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022402.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022403.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022404.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022408.rn.1 | TC0800002755.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800022409.rn.1 | TC0800002755.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376085_PM_at | - | - |
| Rat Expression Set 230 | 1376085_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10935782 | - | - |
| Rat Genome U34 Set | rc_AA849731_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21535537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374026 | - | - |

Tmem185a
Genomic Map