Assay Details
Gene Symbol: Clca5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107450.1 | NP_001100920.1 | 13 - 14 | 2580 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P147300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167869 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P147300 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P147300 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P147300 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P147300 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10705 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10827217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827218 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827220 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10827216 | - | - |
Rat Genome 230 2.0 Array | 1393147_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393147_at | - | - |
Affymetrix HT Rat Focus Array | 1393147_at | - | - |
Affymetrix HT Rat Focus Array | 1393147_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200033599.rn.1 | TC0200004854.rn.1 | RTA TC |
Rat Expression Set 230 | 1393147_at | - | - |
Rat Expression Set 230 | 1393147_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10827216 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21850080 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361466 | - | - |
