Assay Details
| Gene Symbol: Pifo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001077265.5 | XP_001077265.2 | 4 - 5 | 575 | 108 | |
| XM_003749376.3 | XP_003749424.1 | 4 - 5 | 586 | 108 | ||
More Information
Set Membership: |
|

Pifo
Genomic Map