Assay Details
| Gene Symbol: LOC685067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749409.3 | XP_003749457.1 | 2 - 3 | 402 | 68 | |
| GenBank mRNA | FQ221477.1 | - | 308 | 68 | ||
| FQ222982.1 | - | 2 - 3 | 291 | 68 | ||
More Information
Set Membership: |
|

LOC685067
Genomic Map