Assay Details
| Gene Symbol: LOC685067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749409.3 | XP_003749457.1 | 7 - 8 | 1342 | 95 | |
| GenBank mRNA | FQ221477.1 | - | 1266 | 95 | ||
| FQ222982.1 | - | 7 - 8 | 1249 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC685067
Genomic Map