Assay Details
| Gene Symbol: Gipc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037210.1 | NP_001032287.1 | 4 - 5 | 733 | 78 | |
| XM_008761552.1 | XP_008759774.1 | 4 - 5 | 1959 | 78 | ||
| GenBank mRNA | BC100257.1 | - | 4 - 5 | 733 | 7442312 | 78 |
More Information
Updated Mapping Information
Set Membership: |
|

Gipc2
Genomic Map