Assay Details
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019359.1 | NP_062232.1 | 1 - 2 | 293 | 53 | |
GenBank mRNA | BC062020.1 | - | 1 - 2 | 296 | 5623281 | 53 |
U06755.1 | - | 1 - 2 | 293 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
