Assay Details
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019359.1 | NP_062232.1 | 3 - 4 | 476 | 140 | |
GenBank mRNA | BC062020.1 | - | 3 - 4 | 479 | 5623281 | 140 |
U06755.1 | - | 3 - 4 | 476 | 140 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
