Assay Details
| Gene Symbol: Prpf38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024305.2 | NP_001019476.1 | 3 - 4 | 770 | 104 | |
| XM_006233172.1 | XP_006233234.1 | 4 - 5 | 526 | 104 | ||
| XM_008761421.1 | XP_008759643.1 | 3 - 4 | 821 | 104 | ||
| GenBank mRNA | BC079264.1 | - | 3 - 4 | 719 | 7133560 | 104 |
| FQ210223.1 | - | 3 - 4 | 750 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prpf38b
Genomic Map