Assay Details
| Gene Symbol: Ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019276.3 | NP_062149.1 | 5 - 6 | 1330 | 91 | |
| GenBank mRNA | L21698.1 | - | 5 - 6 | 1377 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein localization to paranode region of axon
galactosylceramide biosynthetic process
glycosphingolipid biosynthetic process
cytoskeleton organization
axo-dendritic transport
flavonoid biosynthetic process
paranodal junction assembly
cellular response to hormone stimulus
myelination
neuron projection morphogenesis
flavonoid glucuronidation
2-hydroxyacylsphingosine 1-beta-galactosyltransferase activity
UDP-galactose:glucosylceramide beta-1,4-galactosyltransferase activity
glucuronosyltransferase activity
protein localization to paranode region of axon
galactosylceramide biosynthetic process
glycosphingolipid biosynthetic process
cytoskeleton organization
axo-dendritic transport
flavonoid biosynthetic process
paranodal junction assembly
cellular response to hormone stimulus
myelination
neuron projection morphogenesis
flavonoid glucuronidation
2-hydroxyacylsphingosine 1-beta-galactosyltransferase activity
UDP-galactose:glucosylceramide beta-1,4-galactosyltransferase activity
glucuronosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P047221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30716 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047221 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P047221 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P047221 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12003 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85574 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36360 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36846 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1562 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10826609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826614 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826613 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10826607 | - | - |
| Rat Genome 230 2.0 Array | 1368858_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368858_at | - | - |
| Rat Genome 230 2.0 Array | 1368857_at | - | - |
| Rat Genome 230 2.0 Array | 1368857_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368858_at | - | - |
| Affymetrix HT Rat Focus Array | 1368857_at | - | - |
| Affymetrix HT Rat Focus Array | 1368858_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368857_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200032199.rn.1 | TC0200004631.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368858_PM_at | - | - |
| Rat Expression Set 230 | 1368857_PM_at | - | - |
| Rat Expression Set 230 | 1368858_at | - | - |
| Rat Expression Set 230 | 1368857_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826607 | - | - |
| Rat Genome U34 Set | U07683_at | - | - |
| Rat Genome U34 Set | rc_AI228110_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21988733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650105 | - | - |

Ugt8
Genomic Map