Assay Details
| Gene Symbol: Atp5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134365.2 | NP_599192.1 | 1 - 2 | 185 | 61 | |
| GenBank mRNA | BC063808.1 | - | 1 - 2 | 126 | 6920021 | 61 |
| FQ209675.1 | - | 1 - 2 | 84 | 61 | ||
| FQ212544.1 | - | 1 - 2 | 121 | 61 | ||
| FQ214353.1 | - | 1 - 2 | 147 | 61 | ||
| FQ214371.1 | - | 1 - 2 | 121 | 61 | ||
| FQ214449.1 | - | 1 - 2 | 84 | 61 | ||
| FQ214672.1 | - | 1 - 2 | 120 | 61 | ||
| FQ214784.1 | - | 1 - 2 | 119 | 61 | ||
| FQ215244.1 | - | 1 - 2 | 84 | 61 | ||
| FQ215436.1 | - | 1 - 2 | 84 | 61 | ||
| FQ215480.1 | - | 1 - 2 | 84 | 61 | ||
| FQ215788.1 | - | 1 - 2 | 120 | 61 | ||
| FQ215823.1 | - | 1 - 2 | 137 | 61 | ||
| FQ216062.1 | - | 1 - 2 | 86 | 61 | ||
| FQ216247.1 | - | 1 - 2 | 70 | 61 | ||
| FQ216597.1 | - | 1 - 2 | 76 | 61 | ||
| FQ216773.1 | - | 1 - 2 | 84 | 61 | ||
| FQ216829.1 | - | 1 - 2 | 84 | 61 | ||
| FQ216922.1 | - | 1 - 2 | 84 | 61 | ||
| FQ219899.1 | - | 1 - 2 | 113 | 61 | ||
| FQ223652.1 | - | 1 - 2 | 84 | 61 | ||
| FQ223683.1 | - | 1 - 2 | 110 | 61 | ||
| FQ229261.1 | - | 1 - 2 | 144 | 61 | ||
| FQ229976.1 | - | 1 - 2 | 121 | 61 | ||
| FQ232356.1 | - | 1 - 2 | 121 | 61 | ||
| M35052.1 | - | 1 - 2 | 104 | 61 | ||
| Gene Symbol: LOC100911417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749372.2 | XP_003749420.1 | 180 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5f1
Genomic Map