Assay Details
| Gene Symbol: Atp5f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134365.2 | NP_599192.1 | 4 - 5 | 522 | 159 | |
| GenBank mRNA | BC063808.1 | - | 4 - 5 | 463 | 6920021 | 159 |
| FQ209675.1 | - | 4 - 5 | 421 | 159 | ||
| FQ212544.1 | - | 4 - 5 | 458 | 159 | ||
| FQ214336.1 | - | 4 - 5 | 450 | 159 | ||
| FQ214353.1 | - | 4 - 5 | 484 | 159 | ||
| FQ214371.1 | - | 4 - 5 | 458 | 159 | ||
| FQ214449.1 | - | 4 - 5 | 421 | 159 | ||
| FQ214672.1 | - | 4 - 5 | 457 | 159 | ||
| FQ214784.1 | - | 4 - 5 | 456 | 159 | ||
| FQ215244.1 | - | 4 - 5 | 421 | 159 | ||
| FQ215436.1 | - | 4 - 5 | 421 | 159 | ||
| FQ215480.1 | - | 4 - 5 | 421 | 159 | ||
| FQ215788.1 | - | 4 - 5 | 457 | 159 | ||
| FQ215823.1 | - | 4 - 5 | 474 | 159 | ||
| FQ216062.1 | - | 4 - 5 | 423 | 159 | ||
| FQ216247.1 | - | 4 - 5 | 407 | 159 | ||
| FQ216597.1 | - | 4 - 5 | 413 | 159 | ||
| FQ216773.1 | - | 4 - 5 | 421 | 159 | ||
| FQ216829.1 | - | 4 - 5 | 421 | 159 | ||
| FQ218701.1 | - | 4 - 5 | 457 | 159 | ||
| FQ219899.1 | - | 4 - 5 | 450 | 159 | ||
| FQ223683.1 | - | 4 - 5 | 447 | 159 | ||
| FQ229261.1 | - | 4 - 5 | 481 | 159 | ||
| FQ229976.1 | - | 4 - 5 | 458 | 159 | ||
| FQ232356.1 | - | 4 - 5 | 458 | 159 | ||
| M35052.1 | - | 4 - 5 | 441 | 159 | ||
| Gene Symbol: LOC100911417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749372.2 | XP_003749420.1 | 517 | 159 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5f1
Genomic Map