Assay Details
| Gene Symbol: Srsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035255.1 | NP_001030332.1 | 8 - 9 | 1013 | 84 | |
| XM_006233540.2 | XP_006233602.1 | 9 - 10 | 1053 | 84 | ||
| XM_006233541.2 | XP_006233603.1 | 9 - 10 | 1052 | 84 | ||
| XM_006233542.2 | XP_006233604.1 | 9 - 10 | 1053 | 84 | ||
| XM_006233543.2 | XP_006233605.1 | 10 - 11 | 1110 | 84 | ||
| XM_008761567.1 | XP_008759789.1 | 9 - 10 | 1054 | 84 | ||
| XM_008761568.1 | XP_008759790.1 | 10 - 11 | 4101 | 84 | ||
| XM_008761569.1 | XP_008759791.1 | 10 - 11 | 1117 | 84 | ||
| XM_008761570.1 | XP_008759792.1 | 6 - 7 | 1301 | 84 | ||
| GenBank mRNA | BC099157.1 | - | 8 - 9 | 1013 | 7381485 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Srsf11
Genomic Map