Assay Details
| Gene Symbol: Fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233519.2 | XP_006233581.1 | 1 - 2 | 105 | 73 | |
| GenBank mRNA | BC088089.1 | - | 1 - 2 | 81 | 73 | |
More Information
Set Membership: |
|

Fpgt
Genomic Map