Assay Details
| Gene Symbol: Amy1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001010970.1 | NP_001010970.1 | 2 - 3 | 249 | 114 | |
| GenBank mRNA | AB057450.1 | - | 2 - 3 | 249 | 114 | |
| BC088228.1 | - | 1 - 2 | 192 | 7323054 | 114 | |
| FQ219213.1 | - | 2 - 3 | 578 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Amy1a
Genomic Map