Assay Details
| Gene Symbol: LOC102549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761485.1 | XP_008759707.1 | 4 - 5 | 603 | 85 | |
| Gene Symbol: Elovl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134383.2 | NP_599210.1 | 4 - 5 | 460 | 85 | |
| GenBank mRNA | AB071986.1 | - | 4 - 5 | 460 | 85 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P836392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P428691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339011 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2652 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P836392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P428691 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P836392 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P428691 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P836392 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P428691 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P836392 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P428691 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83871 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57525 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78975 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68975 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43531 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96971 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15623 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78090 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53790 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6063 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14269 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10819006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819011 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819008 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10819005 | - | - |
| Rat Genome 230 2.0 Array | 1394401_at | - | - |
| Rat Genome 230 2.0 Array | 1372318_at | - | - |
| Rat Genome 230 2.0 Array | 1394401_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388108_at | - | - |
| Rat Genome 230 2.0 Array | 1388108_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372318_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388108_at | - | - |
| Affymetrix HT Rat Focus Array | 1394401_at | - | - |
| Affymetrix HT Rat Focus Array | 1388108_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394401_PM_at | - | - |
| Rat Expression Set 230 | 1388108_PM_at | - | - |
| Rat Expression Set 230 | 1388108_at | - | - |
| Rat Expression Set 230 | 1394401_at | - | - |
| Rat Expression Set 230 | 1394401_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819005 | - | - |
| Rat Genome U34 Set | rc_AI145640_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22012416 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367680 | - | - |
| RatRef-12 Expression BeadChip | ILMN_56417 | - | - |

LOC102549542
Genomic Map