Assay Details
| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106471.1 | NP_001099941.1 | 8 - 9 | 1168 | 62 | |
| GenBank mRNA | BC166793.1 | - | 8 - 9 | 1241 | 7937610 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Papss1
Genomic Map