Assay Details
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100518.1 | NP_001093988.1 | 5 - 6 | 963 | 72 | |
XM_008761495.1 | XP_008759717.1 | 5 - 6 | 1019 | 72 | ||
GenBank mRNA | AY724508.1 | - | 5 - 6 | 963 | 72 | |
BC091291.1 | - | 3 - 4 | 319 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
