Assay Details
Gene Symbol: Ndst4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191849.1 | NP_001178778.1 | 9 - 10 | 2122 | 59 | |
XM_006233274.2 | XP_006233336.1 | 10 - 11 | 2443 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
