Assay Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191716.1 | NP_001178645.1 | 4 - 5 | 1562 | 63 | |
| XM_006233268.2 | XP_006233330.1 | 5 - 6 | 2233 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndst3
Genomic Map