Assay Details
| Gene Symbol: LOC100363178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761438.1 | XP_008759660.1 | 3 - 4 | 887 | 69 | |
| XM_008761439.1 | XP_008759661.1 | 3 - 4 | 887 | 69 | ||
| XM_008761440.1 | XP_008759662.1 | 3 - 4 | 887 | 69 | ||
| XM_008761441.1 | XP_008759663.1 | 2 - 3 | 811 | 69 | ||
| XM_008761442.1 | XP_008759664.1 | 2 - 3 | 350 | 69 | ||
| XM_008761443.1 | XP_008759665.1 | 2 - 3 | 294 | 69 | ||
| XM_008761444.1 | XP_008759666.1 | 2 - 3 | 132 | 69 | ||
| GenBank mRNA | BC097395.1 | - | 153 | 7453330 | 69 | |
| FM077301.1 | - | 2 - 3 | 306 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100363178
Genomic Map