Assay Details
Gene Symbol: Tmem56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135879.1 | NP_001129351.1 | 2 - 3 | 242 | 125 | |
XM_006233244.2 | XP_006233306.1 | 4 - 5 | 516 | 125 | ||
XM_006233246.2 | XP_006233308.1 | 4 - 5 | 519 | 125 | ||
XM_006233251.2 | XP_006233313.1 | 4 - 5 | 339 | 125 | ||
XM_006233252.2 | XP_006233314.1 | 3 - 4 | 287 | 125 | ||
XM_006233253.2 | XP_006233315.1 | 3 - 4 | 421 | 125 | ||
XM_006233254.2 | XP_006233316.1 | 3 - 4 | 424 | 125 | ||
XM_008761467.1 | XP_008759689.1 | 3 - 4 | 686 | 125 | ||
XM_008761468.1 | XP_008759690.1 | 3 - 4 | 320 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
