Assay Details
| Gene Symbol: Tspan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004090.2 | NP_001004090.2 | 7 - 8 | 743 | 99 | |
| GenBank mRNA | AF540877.1 | - | 7 - 8 | 769 | 99 | |
| BC100135.1 | - | 7 - 8 | 743 | 7315329 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P973562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89992 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86642 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113481 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P973562 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P973562 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P973562 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233736 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233742 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P973562 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33870 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60337 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38274 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30864 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16373 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33535 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37395 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79899 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40574 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13829 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25023 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10819443 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819448 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819444 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819450 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819447 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819449 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819446 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10819442 | - | - |
| Rat Genome 230 2.0 Array | 1376647_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377089_a_at | - | - |
| Rat Genome 230 2.0 Array | 1394316_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1380725_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1380725_at | - | - |
| Rat Genome 230 2.0 Array | 1376647_at | - | - |
| Rat Genome 230 2.0 Array | 1394316_a_at | - | - |
| Rat Genome 230 2.0 Array | 1377089_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1380725_at | - | - |
| Affymetrix HT Rat Focus Array | 1377089_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1380725_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1377089_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1394316_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1394316_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200016095.rn.1 | TC0200002302.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376647_at | - | - |
| Rat Expression Set 230 | 1377089_a_at | - | - |
| Rat Expression Set 230 | 1394316_PM_a_at | - | - |
| Rat Expression Set 230 | 1376647_PM_at | - | - |
| Rat Expression Set 230 | 1377089_PM_a_at | - | - |
| Rat Expression Set 230 | 1394316_a_at | - | - |
| Rat Expression Set 230 | 1380725_at | - | - |
| Rat Expression Set 230 | 1380725_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10819442 | - | - |
| Rat Genome U34 Set | rc_AA850939_at | - | - |
| Rat Genome U34 Set | rc_AI235665_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20717612 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376901 | - | - |

Tspan5
Genomic Map