Assay Details
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034926.1 | NP_001030098.1 | 2 - 3 | 148 | 69 | |
XM_006233538.2 | XP_006233600.1 | 6 - 7 | 857 | 69 | ||
XM_006233539.2 | XP_006233601.1 | 4 - 5 | 519 | 69 | ||
XM_008761564.1 | XP_008759786.1 | 6 - 7 | 857 | 69 | ||
XM_008761565.1 | XP_008759787.1 | 4 - 5 | 2461 | 69 | ||
XM_008761566.1 | XP_008759788.1 | 5 - 6 | 1639 | 69 | ||
GenBank mRNA | BC105888.1 | - | 2 - 3 | 148 | 7308716 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
