Assay Details
| Gene Symbol: LOC100911379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233104.2 | XP_006233166.1 | 1 - 2 | 934 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006233104.2 transcripts.
Set Membership: |
|

LOC100911379
Genomic Map